missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propranolol Hydrochloride, LoGiCal
Klik for at se tilgængelige muligheder
Quantity:
10 mg
Pakningsstørrelse:
10mg
Tekniske data
Tekniske data
| Chemical Name or Material | Propanolol Hydrochloride |
| CAS | 318-98-9 |
| Analyte or Component Names | Propanolol Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H21 N O2 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-[(1-Methylethyl)amino]-3-(1-naphthalenyloxy)-2-propanol hydrochloride, 1-(Isopropylamino)-3-(1-naphthyloxy)-2-propanol hydrochloride, (+/-)-Propranolol hydrochloride, (R,S)-Propranolol hydrochloride, (RS)-Propranolol hydrochloride, 1-(1-Naphthoxy)-2-hydroxy-3-isopropylaminopropane hydrochloride, 1-(1-Naphthyloxy)-2-hydroxy-3-isopropylaminopropane hydrochloride, 1-(1-Naphthyloxy)-3-(isopropylamino)-2-propanol hydrochloride, 1-(Isopropylamino)-3-(1-naphthoxy)-2-propanol hydrochloride, 1-(Isopropylamino)-3-(1-naphthyloxy)propan-2-ol hydrochloride, Anaprilin, Anapriline, Angilol, Apsolol, Avlocardyl, Bedranol, Beprane, Berkolol, Beta-Neg, Beta-Tablinen, Beta-Timelets, Cardinol, Caridolol, DL-Anapriline, DL-Propranolol hydrochloride, Deralin, Docitan, Dociton, Duranol, Efektolol, Elbol, Frekven, ICI 45520, Inderal, Inderal LA, Indobloc, Intermigran, Kemi S, NSC 91523, Naprilin, Obsidan, Oposim, Prano-Puren, Prophylux, Propranolol chloride, Propranolol hydrochloride, Propranur, Propraratiopharm, Pylapron, Rapynogen, Sagittol, Servanolol, Sloprolol, Sumial, Tesnol, dl-Propranolol hydrochloride |
| InChI Formula | InChI=1S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18H,10-11H2,1-2H3;1H |
| Solution Type | Neat |
| SMILES | Cl.CC(C)NCC(O)COc1cccc2ccccc12 |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion