missing translation for 'onlineSavingsMsg'
Learn More

Propyl Gallate, Mikromol™

Product Code. 17827911
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17827911 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17827911 Supplier Mikromol Supplier No. MM1037.000250
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Propyl Gallate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gallic acid-propyl ester
CAS 121-79-9
Analyte or Component Names Gallic acid-propyl ester
Shipping Condition Room Temperature
Molecular Formula C10 H12 O5
Certifications/Compliance ISO 17034
Synonym Propyl Gallate, 3,4,5-Trihydroxybenzoic acid propyl ester, Gallic acid propyl ester, Antioxidant PG, E 310, Gallic acid n-propyl ester, Hemostyptin, Marupi Gallate, NSC 2626, Nipa 49, Nipagallin P, Nipanox S 1, PG, Progallin P, Propyl 3,4,5-trihydroxybenzoate, Propyl gallate, Tenox PG, n-Propyl 3,4,5-trihydroxybenzoate, n-Propyl gallate
InChI Formula InChI=1S/C10H12O5/c1-2-3-15-10(14)6-4-7(11)9(13)8(12)5-6/h4-5,11-13H,2-3H2,1H3
SMILES CCCOC(=O)c1cc(O)c(O)c(O)c1
IUPAC Name propyl 3,4,5-trihydroxybenzoate
Molecular Weight (g/mol) 212.2
Quantity 250 mg
Formula Weight 212.0685 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.