missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prosurfactant Protein C Antibody (5E6A9) - BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Prosurfactant Protein C Monoclonal specifically detects Prosurfactant Protein C in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | Prosurfactant Protein C |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 5E6A9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | PSP-C, pulmonary surfactant-associated protein C, Pulmonary surfactant-associated proteolipid SPL(Val), SFTP2, SFTP2pulmonary surfactant apoprotein-2 SP-C, SMDP2surfactant, pulmonary-associated protein C, SP-CSP5, surfactant protein C |
| Gene Symbols | SFTPC |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?