missing translation for 'onlineSavingsMsg'
Learn More

Prosurfactant Protein C Antibody (5E6A9) - BSA Free, Novus Biologicals™

Product Code. 18651836 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18651836 0.025 mL 0.03mL
18121158 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18651836 Supplier Novus Biologicals Supplier No. NBP2374250.025ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Prosurfactant Protein C Monoclonal specifically detects Prosurfactant Protein C in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prosurfactant Protein C
Applications Western Blot, ELISA
Classification Monoclonal
Clone 5E6A9
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, ELISA 1:10000
Formulation PBS with 0.05% Sodium Azide
Gene Alias PSP-C, pulmonary surfactant-associated protein C, Pulmonary surfactant-associated proteolipid SPL(Val), SFTP2, SFTP2pulmonary surfactant apoprotein-2 SP-C, SMDP2surfactant, pulmonary-associated protein C, SP-CSP5, surfactant protein C
Gene Symbols SFTPC
Host Species Mouse
Immunogen Purified recombinant fragment of human SFTPC (AA: 60-180 ) expressed in E. Coli.
Molecular Weight of Antigen 21 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6440
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.