missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteasome 19S S3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94013-0.1ml
This item is not returnable.
View return policy
Description
Proteasome 19S S3 Polyclonal antibody specifically detects Proteasome 19S S3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Proteasome 19S S3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| 26S proteasome regulatory subunit RPN3, 26S proteasome regulatory subunit S3, P58, proteasome (prosome, macropain) 26S subunit, non-ATPase, 3, Proteasome subunit p58, RPN3, S3, 26S proteasome non-ATPase regulatory subunit 3, tissue specific transplantation antigen 2, TSTA2 | |
| A synthetic peptide corresponding to a sequence within amino acids 700-800 of human Proteasome 19S S3 (O43242). | |
| 0.1 mL | |
| Neuroscience, Ubiquitin Proteasome Pathway | |
| 5709 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction