missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteasome 19S S3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
188.00 € - 470.00 €
Specifications
| Antigen | Proteasome 19S S3 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18658941
|
Novus Biologicals
NBP2-94013-0.02ml |
0.02 mL |
188.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18686102
|
Novus Biologicals
NBP2-94013-0.1ml |
0.1 mL |
470.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Proteasome 19S S3 Polyclonal antibody specifically detects Proteasome 19S S3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Proteasome 19S S3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience, Ubiquitin Proteasome Pathway | |
| PBS (pH 7.3), 50% glycerol | |
| 5709 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 26S proteasome regulatory subunit RPN3, 26S proteasome regulatory subunit S3, P58, proteasome (prosome, macropain) 26S subunit, non-ATPase, 3, Proteasome subunit p58, RPN3, S3, 26S proteasome non-ATPase regulatory subunit 3, tissue specific transplantation antigen 2, TSTA2 | |
| A synthetic peptide corresponding to a sequence within amino acids 700-800 of human Proteasome 19S S3 (O43242). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title