missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Protein Phosphatase 1 beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30766-25ug
This item is not returnable.
View return policy
Description
Protein Phosphatase 1 beta Polyclonal antibody specifically detects Protein Phosphatase 1 beta in Bovine,Human,Mouse,Rat samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Protein Phosphatase 1 beta | |
| Polyclonal | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Bovine, Human, Mouse, Rat | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| EC 3.1.3.16, EC 3.1.3.53, MGC3672, PP1beta, PP-1Bprotein phosphatase 1, catalytic subunit, delta isoform, PPP1CD, protein phosphatase 1, catalytic subunit, beta isoform, protein phosphatase 1, catalytic subunit, beta isozyme, protein phosphatase 1-beta, protein phosphatase 1-delta, serine/threonine protein phosphatase PP1-beta catalytic subunit, serine/threonine-protein phosphatase PP1-beta catalytic subunit | |
| The immunogen used to generate the purified antibody was a peptide conjugated to KLH corresponding to the sequence NH2-Pro-Arg-Thr-Ala-Asn-Pro-Pro-Lys-LysArg-COOH. This peptide antibody corresponds to C -terminal peptide of Protein Phosphatase 1 beta catalytic subunit having a MW of 38kDa. The sequence used is amino acid 318-327. | |
| 25 μg | |
| Cancer, Lipid and Metabolism, Protein Phosphatase | |
| 5500 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction