missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Protein Phosphatase 1 beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 508.00 €
Specifications
| Antigen | Protein Phosphatase 1 beta |
|---|---|
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066984
|
Novus Biologicals™
NBP3-30766-100ug |
100 μg |
508.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066824
|
Novus Biologicals™
NBP3-30766-25ug |
25 μg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Protein Phosphatase 1 beta Polyclonal antibody specifically detects Protein Phosphatase 1 beta in Bovine,Human,Mouse,Rat samples. It is validated for Immunoprecipitation,Western BlotSpecifications
| Protein Phosphatase 1 beta | |
| Polyclonal | |
| Purified | |
| RUO | |
| Bovine, Human, Mouse, Rat | |
| EC 3.1.3.16, EC 3.1.3.53, MGC3672, PP1beta, PP-1Bprotein phosphatase 1, catalytic subunit, delta isoform, PPP1CD, protein phosphatase 1, catalytic subunit, beta isoform, protein phosphatase 1, catalytic subunit, beta isozyme, protein phosphatase 1-beta, protein phosphatase 1-delta, serine/threonine protein phosphatase PP1-beta catalytic subunit, serine/threonine-protein phosphatase PP1-beta catalytic subunit | |
| The immunogen used to generate the purified antibody was a peptide conjugated to KLH corresponding to the sequence NH2-Pro-Arg-Thr-Ala-Asn-Pro-Pro-Lys-LysArg-COOH. This peptide antibody corresponds to C -terminal peptide of Protein Phosphatase 1 beta catalytic subunit having a MW of 38kDa. The sequence used is amino acid 318-327. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Lipid and Metabolism, Protein Phosphatase | |
| Phosphate buffered saline | |
| 5500 | |
| IgG | |
| Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title