missing translation for 'onlineSavingsMsg'
Learn More

PRR5L Antibody, DyLight 680, Novus Biologicals™

Product Code. 18252678 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18252678 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18252678 Supplier Novus Biologicals Supplier No. NBP282025FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PRR5L Polyclonal antibody specifically detects PRR5L in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRR5L
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias FLJ14213MGC16218, FLJ22630, proline rich 5 like, Protein observed with Rictor-2, protor-2, PROTOR2proline-rich protein 5-like
Host Species Rabbit
Immunogen PRR5L antibody was raised against a 17 amino acid synthetic peptide near the center of human PRR5L. The immunogen is located within amino acids 220 - 270 of PRR5L. Amino Acid Squence: SGPTYTLARRHSRVRPKV
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 79899
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.