missing translation for 'onlineSavingsMsg'
Learn More

PRRX1 Antibody (CL14471) [mFluor Violet 610 SE], Novus Biologicals Biologicals™

Product Code. 30532012 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30532012 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30532012 Supplier Novus Biologicals Supplier No. NBP344310MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PRRX1 Monoclonal antibody specifically detects PRRX1 in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRRX1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone CL14471
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias Homeobox protein PHOX1, paired mesoderm homeo box 1, paired mesoderm homeobox 1 isoform pmx-1b, paired mesoderm homeobox protein 1, paired related homeobox 1, Paired-related homeobox protein 1, PHOX1PRX-1, PMX1PRX1
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of P54821, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5396
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.