Learn More
PSMB8 Rabbit, anti-Human, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL594-14859-20UL
 View more versions of this product
Product Code. 18076405
  • 140.00 € / 20µL
Description
The PSMB8 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PSMB8. This antibody recognizes human, mouse antigen. The PSMB8 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
Specifications
PSMB8
1000 μg/mL
beta5i, D6S216, D6S216E, LMP7, Low molecular mass protein 7, Macropain subunit C13, Proteasome component C13, Proteasome subunit beta 5i, Proteasome subunit beta type 8, PSMB5i, PSMB8, RING10, Y2
20 μL
5696
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite 594
Rabbit
Primary
Human, Mouse
150 kDa