Learn More
Invitrogen™ PTP1B/PTPN2 Recombinant Rabbit Monoclonal Antibody (24GB1130)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556941
 View more versions of this product
Product Code. 30274792
  • 146.00 € / 20µL
Description
PTP1B/PTPN2 Recombinant Monoclonal Antibody for Western Blot, Flow
PTP1B (Protein Tyrosine Phosphatase 1B), encoded by the PTPN1 gene located on chromosome 20q13.1, is a major regulator of several cellular signaling pathways, impacting diseases like diabetes, obesity, and various cancers. The protein structure includes multiple exons and functions as a non-transmembrane enzyme that negatively regulates signaling cascades, including insulin and leptin signaling, which are vital for metabolic homeostasis. PTPN2 (Protein Tyrosine Phosphatase Non-Receptor Type 2), located on chromosome 18p11.3-p11.2, also known as T-cell PTP, is similarly a non-transmembrane protein tyrosine phosphatase that modulates immune responses and cell growth. It has ten exons and produces two splice variants, impacting signaling pathways by regulating the phosphorylation state of key proteins. Both of these phosphatases are involved in the fine-tuning of signaling mechanisms that control cell growth, differentiation, and immune regulation, highlighting their importance in disease pathology and as potential therapeutic targets.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PTP1B/PTPN2
Recombinant Monoclonal
0.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P17706, P18031, P20417, P35233, P35821, Q06180
PTPN1, PTPN2
A synthesized peptide derived from human PTP1B/PTPN2 (aa 11-66).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1130
Unconjugated
PTPN1
protein tyrosine phosphatase 1B; PTP1B; T-cell protein tyrosine phosphatase; TCPTP
Rabbit
Affinity chromatography
RUO
117063, 19246, 19255, 24697, 5770, 5771
-20°C