missing translation for 'onlineSavingsMsg'
Learn More

PTPRK Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18647150 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18647150 0.1 mL 0.1mL
18693221 0.02 mL 0.02mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18647150 Supplier Novus Biologicals Supplier No. NBP2938410.1ml

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PTPRK Polyclonal antibody specifically detects PTPRK in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRK
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias DKFZp686C2268, DKFZp779N1045, EC 3.1.3.48, protein tyrosine phosphatase kappa, protein tyrosine phosphatase kappa, protein tyrosine phosphatase, receptor type, K, Protein-tyrosine phosphatase kappa, protein-tyrosine phosphatase, receptor type, kappa, PTPK, receptor type, K (R-PTP-KAPPA, receptor-type tyrosine-protein phosphatase kappa
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 600-750 of human PTPRK (NP_001129120.1). EGVDASLNETATTITVLLRPAQAKGAPISAYQIVVEELHPHRTKREAGAMECYQVPVTYQNAMSGGAPYYFAAELPPGNLPEPAPFTVGDNRTYQGFWNPPLAPRKGYNIYFQAMSSVEKETKTQCVRIATKAAATEEPEVIPDPAKQTDR
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 5796
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.