missing translation for 'onlineSavingsMsg'
Learn More

PX19 Antibody - BSA Free, Novus Biologicals™

Product Code. 30557310 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30557310 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30557310 Supplier Novus Biologicals Supplier No. NBP346900

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PX19 Polyclonal antibody specifically detects PX19 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen PX19
Applications ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:20-1:200
Formulation PBS, 50% Glycerol, pH (7.3)
Gene Alias 25 kDa protein of relevant evolutionary and lymphoid interest, CGI-106, MGC87972, PRELI domain containing 1, PRELImitochondrial, protein of relevant evolutionary and lymphoid interest
Host Species Rabbit
Immunogen PRELI domain containing 1. (Uniprot# Q9Y255)
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27166
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.