missing translation for 'onlineSavingsMsg'
Learn More

Pyridostigmine Bromide, LoGiCal

Product Code. 30462804
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30462804 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462804 Supplier LoGiCal Supplier No. LGCFOR0429.00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material Pyridostigmine Bromide
CAS 101-26-8
Analyte or Component Names Pyridostigmine Bromide
Shipping Condition Room Temperature
Molecular Formula C9 H13 N2 O2 . Br
Certifications/Compliance ISO 9001
Synonym Pyridostigmine Bromide, 3-Hydroxy-1-methylpyridinium bromide dimethylcarbamate, Dimethylcarbamic acid ester with 3-hydroxy-1-methylpyridinium bromide, 1-Methyl-3-hydroxypyridinium bromide dimethylcarbamate, 3-(Dimethylcarbamoyloxy)-1-methylpyridinium bromide, 3-(Dimethylcarbamyloxy)-1-methylpyridinium bromide, 3-[[(Dimethylamino)carbonyl]oxy]-1-methylpyridinium bromide, Kalimin, Kalymin, Mestinon, Mestinon bromide, NSC 679759, Piridostigmin, Piridostigmin bromide, Pyridostigmine bromide, Regonol, Ro 1-5130
InChI Formula InChI=1S/C9H13N2O2.BrH/c1-10(2)9(12)13-8-5-4-6-11(3)7-8;/h4-7H,1-3H3;1H/q+1;/p-1
Solution Type Neat
SMILES [Br-].CN(C)C(=O)Oc1ccc[n+](C)c1
IUPAC Name (1-methylpyridin-1-ium-3-yl) N,N-dimethylcarbamate;bromide
Industry Type Clinical Testing
Molecular Weight (g/mol) 261.12
Quantity 10 mg
Formula Weight 260.016 g/mol
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.