missing translation for 'onlineSavingsMsg'
Learn More

Pyrimidine-2,4,6-triamine, Mikromol™

Product Code. 17904596
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17904596 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17904596 Supplier Mikromol Supplier No. MM0269.070025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Pyrimidine-2,4,6-triamine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pyrimidine-2,4,6-triamine
CAS 1004-38-2
Analyte or Component Names Pyrimidine-2,4,6-triamine
Shipping Condition Room Temperature
Molecular Formula C4 H7 N5
Certifications/Compliance ISO 9001
Synonym 2,4,6-Pyrimidinetriamine, Pyrimidine, 2,4,6-triamino- (6CI,7CI,8CI), 2,4,6-Triaminopyrimidine, NSC 26493, Pyrimidine-2,4,6-triamine
InChI Formula InChI=1S/C4H7N5/c5-2-1-3(6)9-4(7)8-2/h1H,(H6,5,6,7,8,9)
SMILES Nc1cc(N)nc(N)n1
IUPAC Name pyrimidine-2,4,6-triamine
Molecular Weight (g/mol) 125.07
Quantity 25 mg
Formula Weight 125.132 g/mol
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.