Learn More
Invitrogen™ Qualification Add-on For FL1000 Imaging System
Description
- Verify instrument performance and minimize downtime with IQ/OQ services (Installation Qualification and Operation Qualification)
- Complete with audit ready documentation, factory-trained and experienced engineers will conduct IQ/OQ services
- Qualification services help ensure instruments are installed and operating according to manufacturer's specifications
- Helps move into production faster
- IQ/OQ recommended: At installation , after a move
- OQ recommended: After planned maintenance, After major repair, After additions, changes, or upgrades to the system, For a scheduled OQ site requirement
Spezifikation
Spezifikation
| For Use With (Equipment) | iBright FL1000 Imaging System |
| Product Type | Qualification Add-on |
| For Use With (Application) | Warranty for Western Blotting instrumentation |
| Includes | Audit-ready documentation, Order and system verification, Documentation verification, Installation verification, Software verification, Hardware operation verification, Component verification, Service coverage verification, Customer training verification |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?