missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quetiapine Fumarate EP Impurity F (as Maleate), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Quetiapine Fumarate EP Impurity F (as Maleate)
Specifications
Specifications
| Chemical Name or Material | (2-[(2-Aminophenyl)thio]phenyl][4-[2-(2-hydroxyethoxy)ethyl]piperazin-1-yl)methanone Maleate |
| Analyte or Component Names | (2-[(2-Aminophenyl)thio]phenyl][4-[2-(2-hydroxyethoxy)ethyl]piperazin-1-yl)methanone Maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H27 N3 O3 S . C4 H4 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | [2-[(2-Aminophenyl)thio]phenyl][4-[2-(2-hydroxyethoxy)ethyl]piperazin-1-yl]methanone Maleate, Quetiapine Fumarate Imp. F (EP), Quetiapine Imp. F (EP), [2-[(2-Aminophenyl)thio]phenyl][4-[2-(2-hydroxyethoxy)ethyl]piperazin-1-yl]methanone Maleate, Quetiapine Fumarate Imp. F (EP) as Maleate, Quetiapine Fumarate Impurity F as Maleate, Quetiapine Impurity F as Maleate |
| InChI Formula | InChI=1S/C19H20F3NO2.ClH/c1-14(13-16-9-5-6-10-17(16)19(20,21)22)23-11-12-25-18(24)15-7-3-2-4-8-15;/h2-10,14,23H,11-13H2,1H3;1H |
| SMILES | Nc1ccccc1Sc2ccccc2C(=O)N3CCN(CCOCCO)CC3.OC(=O)\C=C/C(=O)O |
| IUPAC Name | [2-(2-aminophenyl)sulfanylphenyl]-[4-[2-(2-hydroxyethoxy)ethyl]piperazin-1-yl]methanone;(Z)-but-2-enedioic acid |
| Molecular Weight (g/mol) | 517.59 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction