missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quetiapine Fumarate, Mikromol™
Description
Quetiapine Fumarate
Specifications
Specifications
| Chemical Name or Material | Quetiapine fumarate |
| CAS | 111974-72-2 |
| Analyte or Component Names | Quetiapine fumarate |
| Shipping Condition | Room Temperature |
| Molecular Formula | 2 C21 H25 N3 O2 S . C4 H4 O4 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Quetiapine Fumarate, Ethanol, 2-[2-(4-dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]-, (2E)-2-butenedioate (2:1), Ethanol, 2-[2-(4-dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]-, (2E)-2-butenedioate (2:1) (salt) (9CI), Ethanol, 2-[2-(4-dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]-, (E)-2-butenedioate (2:1) (salt), Dibenzo[b,f][1,4]thiazepine, ethanol deriv., ICI 204636, Quetiapine fumarate, Quetiapine hemifumarate, Seroquel, ZD 5077, ZM 204636 |
| InChI Formula | InChI=1S/2C21H25N3O2S.C4H4O4/c2*25-14-16-26-15-13-23-9-11-24(12-10-23)21-17-5-1-3-7-19(17)27-20-8-4-2-6-18(20)22-21;5-3(6)1-2-4(7)8/h2*1-8,25H,9-16H2;1-2H,(H,5,6)(H,7,8)/b;;2-1+ |
| SMILES | OCCOCCN1CCN(CC1)C2=Nc3ccccc3Sc4ccccc24.OCCOCCN5CCN(CC5)C6=Nc7ccccc7Sc8ccccc68.OC(=O)\C=C\C(=O)O |
| IUPAC Name | 2-[2-(4-benzo[b][1,4]benzothiazepin-6-ylpiperazin-1-yl)ethoxy]ethanol;(E)-but-2-enedioic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?