missing translation for 'onlineSavingsMsg'
Learn More

Quetiapine.hemifumarate, 1mg/ml in Methanol (as free base), Lipomed

Product Code. 30458292
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
30458292 1 mL 1mL
1 options
This item is not returnable. View return policy

Product Code. 30458292

Brand: Lipomed LPMQUE1200FU1LM

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Quetiapine.hemifumarate, 1mg/ml in Methanol (as free base)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Quetiapine fumarate
CAS 111974-72-2
Analyte or Component Names Quetiapine fumarate
Shipping Condition Room Temperature
Molecular Formula 2 C21 H25 N3 O2 S . C4 H4 O4
Concentration 1 mg/mL
Synonym Quetiapine Fumarate, Ethanol, 2-[2-(4-dibenzo[b,f][1,4 ]thiazepin-11-yl-1-piperazinyl)ethoxy]-, (2 E)-2-butenedioate (2:1), Ethanol, 2-[2-(4-dibenzo[b,f][1,4 ]thiazepin-11-yl-1-piperazinyl)ethoxy]-, (2 E)-2-butenedioate (2:1) (salt) (9 CI), Ethanol, 2-[2-(4-dibenzo[b,f][1,4 ]thiazepin-11-yl-1-piperazinyl)ethoxy]-, (E)-2-butenedioate (2:1) (salt), Dibenzo[b,f][1,4 ]thiazepine, ethanol deriv., ICI 204636, Quetiapine fumarate, Quetiapine hemifumarate, Seroquel, ZD 5077, ZM 204636
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES OCCOCCN1CCN(CC1)C2=Nc3ccccc3Sc4ccccc24.OCCOCCN5CCN(CC5)C6=Nc7ccccc7Sc8ccccc68.OC(=O)\C=C\C(=O)O
IUPAC Name 2-[2-(4-benzo[b][1,4 ]benzothiazepin-6-ylpiperazin-1-yl)ethoxy]ethanol;(E)-but-2-enedioic acid
Solvent or Matrix Methanol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 883.09
Quantity 1 mL
Formula Weight 882.3445 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.