missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-2-Methyl-1,2,3,4-tetrahydroisoquinoline-4,8-diol Hydrochloride Monohydrate, Mikromol™
Description
(R)-2-Methyl-1,2,3,4-tetrahydroisoquinoline-4,8-diol Hydrochloride Monohydrate
Specifications
Specifications
| Chemical Name or Material | (R)-2-Methyl-1,2,3,4-tetrahydroisoquinoline-4,8-diol Hydrochloride Monohydrate |
| CAS | 1007885-60-0 |
| Analyte or Component Names | (R)-2-Methyl-1,2,3,4-tetrahydroisoquinoline-4,8-diol Hydrochloride Monohydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H13 N O2 . Cl H . H2 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | 4,8-Isoquinolinediol, 1,2,3,4-tetrahydro-2-methyl-, hydrochloride, hydrate (1:1:1), (4R)-, (R)-2-Methyl-1,2,3,4-tetrahydroisoquinoline-4,8-diol hydrochloride monohydrate, Phenylephrine Related Compound F as Hydrochloride Monohydrate |
| InChI Formula | InChI=1S/C10H13NO2.ClH.H2O/c1-11-5-8-7(10(13)6-11)3-2-4-9(8)12;;/h2-4,10,12-13H,5-6H2,1H3;1H;1H2/t10-;;/m0../s1 |
| SMILES | O.Cl.CN1C[C@H](O)c2cccc(O)c2C1 |
| IUPAC Name | (4R)-2-methyl-3,4-dihydro-1H-isoquinoline-4,8-diol;hydrate;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?