missing translation for 'onlineSavingsMsg'
Learn More

R-Spondin 2 Antibody (CL14321) [Janelia Fluor« 669], Novus Biologicals Biologicals™

Product Code. 30532518 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30532518 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30532518 Supplier Novus Biologicals Supplier No. NBP344331JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

R-Spondin 2 Monoclonal antibody specifically detects R-Spondin 2 in Human,Mouse samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen R-Spondin 2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL14321
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias CRISTIN2, hRspo2, MGC35555, MGC43342, Roof plate-specific spondin-2, R-spondin 2 homolog (Xenopus laevis), R-spondin-2
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of Q6UXX9, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340419
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.