Learn More
Invitrogen™ Rab4 Recombinant Rabbit Monoclonal Antibody (24GB825)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557660
 View more versions of this product
Product Code. 30275265
  • 564.00 € / 100µL
Description
Rab4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Protein transport. Probably involved in vesicular traffic.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rab4
Recombinant Monoclonal
1.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P05714, P20338, P56371
Rab4a
A synthesized peptide derived from human Rab4 (aa 50-75).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB825
Unconjugated
Rab4a
AI848268; HRES1; HRES-1; HRES-1/RAB4; HTLV-1 related endogenous sequence; RAB4; Rab4a; RAB4A, member RAS oncogene family; ras-related protein Rab-4A; Ras-related small GTP binding protein 4
Rabbit
Affinity chromatography
RUO
19341, 25532, 5867
-20°C