missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rab4 Recombinant Rabbit Monoclonal Antibody (24GB825)

Rabbit Recombinant Monoclonal Antibody
198.00 € - 564.00 €
Specifications
| Antigen | Rab4 |
|---|---|
| Clone | 24GB825 |
| Concentration | 1.2 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275265
![]() |
Invitrogen™
MA557660 |
100 μL |
564.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275071
|
Invitrogen™
MA557661 |
20 μL |
198.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rab4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Protein transport. Probably involved in vesicular traffic.Specifications
| Rab4 | |
| 1.2 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P05714, P20338, P56371 | |
| 19341, 25532, 5867 | |
| A synthesized peptide derived from human Rab4 (aa 50-75). | |
| Antibody |
| 24GB825 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| Rab4a | |
| AI848268; HRES1; HRES-1; HRES-1/RAB4; HTLV-1 related endogenous sequence; RAB4; Rab4a; RAB4A, member RAS oncogene family; ras-related protein Rab-4A; Ras-related small GTP binding protein 4 | |
| Rab4a | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title