Learn More
Invitrogen™ RAB5C Recombinant Rabbit Monoclonal Antibody (24GB3875)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557202
 View more versions of this product
Product Code. 30276017
  • 146.00 € / 20µL
Description
RAB5C Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The Ras-related superfamily of guanine nucleotide binding proteins, which includes the R-Ras, Rap, Ral/Rec and Rho/Rab subfamilies, exhibit 30-60% homology with Ras p21. Accumulating data suggests an important role for Rab proteins, either in endocytosis or in biosynthetic protein transport. The transport of newly synthesized proteins from the endoplasmic reticulum to various stacks of the Golgi complex and to secretory vesicles involves at each stage the movement of carrier vesicles, a process that appears to involve Rab protein function. The possibility that Rab proteins might also direct the exocytosis from secretory vesicles to the plasma membrane is supported by the observation that in yeast, the SEC4 protein, which is 40% homologous to Rab proteins, is associated with secretory vesicles.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RAB5C
Recombinant Monoclonal
1.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P35278, P51148
RAB5C
A synthesized peptide derived from human RAB5C / RABL (aa 2-216).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB3875
Unconjugated
RAB5C
AI326010; L1880; RAB; RAB5B; RAB5B, member RAS oncogene family; RAB5C; Rab5c protein; RAB5C, member of RAS oncogene family; RAB5C, member RAS oncogene family; RAB5C, member RAS oncogene family isoform b; RAB5CL; RAB5L; RABL; Ras-related protein Rab-5C; small GTPase; wu:fb98b08; wu:fi16g01
Rabbit
Affinity chromatography
RUO
19345, 287709, 5878
-20°C