missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAB5C Recombinant Rabbit Monoclonal Antibody (24GB3875)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

146.00 € - 500.00 €

Specifications

Antigen RAB5C
Clone 24GB3875
Concentration 1.2 mg/mL
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30276222
GREENER_CHOICE
View Documents
Invitrogen™
MA557201
100 μL
500.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30276017
View Documents
Invitrogen™
MA557202
20 μL
146.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RAB5C Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

The Ras-related superfamily of guanine nucleotide binding proteins, which includes the R-Ras, Rap, Ral/Rec and Rho/Rab subfamilies, exhibit 30-60% homology with Ras p21. Accumulating data suggests an important role for Rab proteins, either in endocytosis or in biosynthetic protein transport. The transport of newly synthesized proteins from the endoplasmic reticulum to various stacks of the Golgi complex and to secretory vesicles involves at each stage the movement of carrier vesicles, a process that appears to involve Rab protein function. The possibility that Rab proteins might also direct the exocytosis from secretory vesicles to the plasma membrane is supported by the observation that in yeast, the SEC4 protein, which is 40% homologous to Rab proteins, is associated with secretory vesicles.
Specifications

Specifications

RAB5C
1.2 mg/mL
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P35278, P51148
19345, 287709, 5878
A synthesized peptide derived from human RAB5C / RABL (aa 2-216).
Antibody
24GB3875
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
RAB5C
AI326010; L1880; RAB; RAB5B; RAB5B, member RAS oncogene family; RAB5C; Rab5c protein; RAB5C, member of RAS oncogene family; RAB5C, member RAS oncogene family; RAB5C, member RAS oncogene family isoform b; RAB5CL; RAB5L; RABL; Ras-related protein Rab-5C; small GTPase; wu:fb98b08; wu:fi16g01
RAB5C
Primary
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.