missing translation for 'onlineSavingsMsg'
Learn More

Rabeprazole Sodium, Mikromol™

Product Code. 17971846
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17971846 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17971846 Supplier Mikromol Supplier No. MM0618.010250
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Rabeprazole Sodium
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Rabeprazole Sodium
CAS 117976-90-6
Analyte or Component Names Rabeprazole Sodium
Shipping Condition Room Temperature
Molecular Formula C18 H20 N3 O3 S . Na
Certifications/Compliance ISO 17034
Synonym 1H-Benzimidazole, 2-[[[4-(3-methoxypropoxy)-3-methyl-2-pyridinyl]methyl]sulfinyl]-, sodium salt (1:1), 1H-Benzimidazole, 2-[[[4-(3-methoxypropoxy)-3-methyl-2-pyridinyl]methyl]sulfinyl]-, sodium salt (9CI), 2-[[[4-(3-Methoxypropoxy)-3-methylpyridin-2-yl]methyl]sulfinyl]-1H-benzimidazole sodium salt, Aciphex, E 3810, LY 307640 sodium, Pariet, Pariprazole, Pepcia, Rabeprazole sodium, Rabicip, Razo, Rabeprazole Sodium Imp. B (EP), Rabeprazole Imp. B (EP), Rabeprazole USP Related Compound E, Rabeprazole USP RC E
InChI Formula InChI=1S/C18H20N3O3S.Na/c1-13-16(19-9-8-17(13)24-11-5-10-23-2)12-25(22)18-20-14-6-3-4-7-15(14)21-18;/h3-4,6-9H,5,10-12H2,1-2H3;/q-1;+1
SMILES [Na+].COCCCOc1ccnc(CS(=O)c2nc3ccccc3[n-]2)c1C
IUPAC Name sodium;2-[[4-(3-methoxypropoxy)-3-methylpyridin-2-yl]methylsulfinyl]benzimidazol-1-ide
Molecular Weight (g/mol) 381.42
Quantity 250 mg
Formula Weight 381.1123 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.