missing translation for 'onlineSavingsMsg'
Learn More

Rabeprazole Sulphone N-Oxide, Mikromol™

Product Code. 30051817
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051817 25 mg 25mg
17923836 100 mg 100mg
30457875 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30051817 Supplier Mikromol Supplier No. MM0618.050025
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Rabeprazole Sulphone N-Oxide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Rabeprazole Sulfone N-Oxide
CAS 924663-37-6
Analyte or Component Names Rabeprazole Sulfone N-Oxide
Shipping Condition Room Temperature
Molecular Formula C18 H21 N3 O5 S
Certifications/Compliance ISO 9001
Synonym Rabeprazole Sulphone N-Oxide, Rabeprazole Sodium Imp. I (EP), Rabeprazole Imp. I (EP), Rabeprazole Sodium Impurity I, Rabeprazole Impurity I
InChI Formula InChI=1S/C18H21N3O5S/c1-13-16(21(22)9-8-17(13)26-11-5-10-25-2)12-27(23,24)18-19-14-6-3-4-7-15(14)20-18/h3-4,6-9H,5,10-12H2,1-2H3,(H,19,20)
SMILES COCCCOc1cc[n+]([O-])c(CS(=O)(=O)c2nc3ccccc3[nH]2)c1C
IUPAC Name 2-[[4-(3-methoxypropoxy)-3-methyl-1-oxidopyridin-1-ium-2-yl]methylsulfonyl]-1H-benzimidazole
Molecular Weight (g/mol) 391.44
Quantity 25 mg
Formula Weight 391.1202 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.