missing translation for 'onlineSavingsMsg'
Learn More

Rabphilin 3A Antibody [DyLight 350], Novus Biologicals Biologicals™

Product Code. 30509525 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30509525 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30509525 Supplier Novus Biologicals Supplier No. NBP338317UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Rabphilin 3A Polyclonal antibody specifically detects Rabphilin 3A in Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabphilin 3A
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias Exophilin-1, KIAA0985exophilin-1, rabphilin, rabphilin 3A homolog (mouse), rabphilin-3A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human Rabphilin 3A (NP_055769.2).,, Sequence:, MTDTVFSNSSNRWMYPSDRPLQSKLQAGWSVHPGGQPDRQRKQEELTDEEKEIINRVIARAEKMEEMEQERIGRLVDRLENMRKNVAGDGVNRCILCGEQLGMLGSACVVCEDCKKNVCTKCGVETNNRLHSVWLCKICIEQREVWKRSGAWFFKGFPKQVLPQPMPIKKTKPQQPVSEPAAPEQPAPEPKHPARAPARGDSEDRRGPGQKTGPDPASAPGRGNYGPPVRRASEARMSSSSRDSESWDHSGGAGDSSRSPAGLRRANSVQASRPAPGSVQ
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 22895
Target Species Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.