missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rac1,2,3 Antibody (SR1332), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21724-100ul
This item is not returnable.
View return policy
Description
Rac1,2,3 Monoclonal antibody specifically detects Rac1,2,3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Rac1,2,3 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| SR1332 | |
| Western Blot 1:500-1:2000 | |
| AI323801;AI452260;AL023026;D5Ertd559e;EN-7;Gx;HSPC022;MIG5;p21-Rac1;p21-Rac2;RAC1;Rac-1;Rac1B;RAC2;RAC3;TC25;TC-25 | |
| A synthesized peptide derived from human Rac1,2,3 (Uniprot #: P63000, P15153, P60763) | |
| 100 μg | |
| B Cell Development and Differentiation Markers, Cancer, Cell Biology, Cytoskeleton Markers, GPCR, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Protein Phosphatase, Signal Transduction | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction