missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rac1,2,3 Antibody (SR1332), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 € - 449.00 €
Specifications
| Antigen | Rac1,2,3 |
|---|---|
| Clone | SR1332 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18681155
|
Novus Biologicals
NBP3-21724-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18690874
|
Novus Biologicals
NBP3-21724-50ul |
50 μg |
338.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rac1,2,3 Monoclonal antibody specifically detects Rac1,2,3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Rac1,2,3 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AI323801;AI452260;AL023026;D5Ertd559e;EN-7;Gx;HSPC022;MIG5;p21-Rac1;p21-Rac2;RAC1;Rac-1;Rac1B;RAC2;RAC3;TC25;TC-25 | |
| IgG | |
| Affinity purified |
| SR1332 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| B Cell Development and Differentiation Markers, Cancer, Cell Biology, Cytoskeleton Markers, GPCR, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Protein Phosphatase, Signal Transduction | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| A synthesized peptide derived from human Rac1,2,3 (Uniprot #: P63000, P15153, P60763) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title