missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ractopamine Antibody (Rac1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28847-0.025ml
This item is not returnable.
View return policy
Description
Ractopamine Monoclonal specifically detects Ractopamine in Non-species specific samples. It is validated for ELISA.
Specifications
| Ractopamine | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| Mouse | |
| Unpurified | |
| RUO | |
| Specific for Ractopamine. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| ELISA | |
| Rac1 | |
| ELISA 1:10000 | |
| C18H23NO3 | |
| Rac-OVA. | |
| 0.025 mL | |
| Primary | |
| Non-species specific | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction