missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ractopamine Antibody (Rac1), Novus Biologicals™
Mouse Monoclonal Antibody
116.00 €
Specifications
| Antigen | Ractopamine |
|---|---|
| Clone | Rac1 |
| Dilution | ELISA 1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
Description
Ractopamine Monoclonal specifically detects Ractopamine in Non-species specific samples. It is validated for ELISA.Specifications
| Ractopamine | |
| ELISA 1:10000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Ascites with 0.03% Sodium Azide | |
| Rac-OVA. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Rac1 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Non-species specific | |
| C18H23NO3 | |
| IgG | |
| Unpurified | |
| Specific for Ractopamine. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title