missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad9 Antibody (93A535) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 8 publications
Brand: Novus Biologicals NB120-13600SS
This item is not returnable.
View return policy
Description
Rad9 Monoclonal specifically detects Rad9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Rad9 | |
| Monoclonal | |
| 1.0 mg/ml | |
| Western Blot 1-2 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (Kotmatsu et al), Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500. Use reported in scientific literature (Lee et al (2007)) | |
| cell cycle checkpoint control protein RAD9A, DNA repair exonuclease rad9 homolog A, EC 3.1.11.2, hRAD9, RAD9, RAD9 (S. pombe) homolog, RAD9 homolog A (S. pombe) | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| In recombinant Rad9, a ∼50 kDa band is observed. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 93A535 | |
| Unconjugated | |
| Q99638 | |
| RAD9A | |
| The antibody was raised against fusion protein of human full length Rad9. | |
| 0.025 mg | |
| Apoptosis, Cancer, Checkpoint signaling, DNA Repair, Tumor Suppressors | |
| 5883 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction