missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad9 Antibody (93A535) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 8 publications
231.00 € - 477.00 €
Specifications
| Antigen | Rad9 |
|---|---|
| Clone | 93A535 |
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (Kotmatsu et al), Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500. Use reported in scientific literature (Lee et al (2007)) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18779693
|
Novus Biologicals
NB120-13600SS |
0.025 mg |
231.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18707063
|
Novus Biologicals
NB120-13600 |
0.1 mg |
477.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rad9 Monoclonal specifically detects Rad9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Rad9 | |
| 1.0 mg/ml | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Checkpoint signaling, DNA Repair, Tumor Suppressors | |
| Q99638 | |
| 5883 | |
| The antibody was raised against fusion protein of human full length Rad9. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 93A535 | |
| Western Blot 1-2 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (Kotmatsu et al), Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500. Use reported in scientific literature (Lee et al (2007)) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cell cycle checkpoint control protein RAD9A, DNA repair exonuclease rad9 homolog A, EC 3.1.11.2, hRAD9, RAD9, RAD9 (S. pombe) homolog, RAD9 homolog A (S. pombe) | |
| RAD9A | |
| IgG1 κ | |
| Protein G purified | |
| In recombinant Rad9, a ∼50 kDa band is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title