missing translation for 'onlineSavingsMsg'
Learn More

RAE1 Antibody [FITC], Novus Biologicals Biologicals™

Product Code. 30509450 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30509450 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30509450 Supplier Novus Biologicals Supplier No. NBP338315F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RAE1 Polyclonal antibody specifically detects RAE1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAE1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias dJ481F12.3, dJ800J21.1, FLJ30608, homolog of yeast Rae1 (Bharathi) mRNA-associated protein of 41 kDa (Kraemer), MGC117333, MGC126076, MGC126077, MIG14, migration-inducing gene 14, Mnrp41, mRNA export factor, mRNA export protein, mRNA-associated protein mrnp 41, mRNA-binding protein, 41-kD, MRNP41, RAE1 (RNA export 1, S.pombe) homolog, Rae1 protein homolog, RAE1 RNA export 1 homolog (S. pombe)
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-368 of human RAE1 (NP_003601.1).,, Sequence:, MSLFGTTSGFGTSGTSMFGSATTDNHNPMKDIEVTSSPDDSIGCLSFSPPTLPGNFLIAGSWANDVRCWEVQDSGQTIPKAQQMHTGPVLDVCWSDDGSKVFTASCDKTAKMWDLSSNQAIQIAQHDAPVKTIHWIKAPNYSCVMTGSWDKTLKFWDTRSSNPMMVLQLPERCYCADVIYPMAVVATAERGLIVYQLENQPSEFRRIESPLKHQHRCVAIFKDKQNKPTGFALGSIEGRVAIHYINPPNPAKDNFTFKCHRSNGTNTSAPQDIYAVNGIAFHPVHGTLATVGSDGRFSFWDKDARTKLKTSEQLDQPISACCFNHNGNIFAYASSYDWSKGHEFYNPQKKNYIFLRNAAEELKPRNKK
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8480
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.