missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RALGDS Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93790-0.02ml
This item is not returnable.
View return policy
Description
RALGDS Polyclonal antibody specifically detects RALGDS in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| RALGDS | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin | |
| ral guanine nucleotide dissociation stimulator, Ral guanine nucleotide exchange factor, RalGDS, RalGEFKIAA1308, RGDS, RGFFLJ20922 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 771-902 of human RALGDS (NP_001258705). CNSSSALPLYNQQVGDCCIIRVSLDVDNGNMYKSILVTSQDKAPAVIRKAMDKHNLEEEEPEDYELLQILSDDRKLKIPENANVFYAMNSTANYDFVLKKRTFTKGVKVKHGASSTLPRMKQKGLKIAKGIF | |
| 0.02 mL | |
| Signal Transduction | |
| 5900 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction