missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RALGDS Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
221.00 € - 470.00 €
Specifications
| Antigen | RALGDS |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18601651
|
Novus Biologicals
NBP2-93790-0.02ml |
0.02 mL |
221.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18617250
|
Novus Biologicals
NBP2-93790-0.1ml |
0.1 mL |
470.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RALGDS Polyclonal antibody specifically detects RALGDS in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| RALGDS | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 5900 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ral guanine nucleotide dissociation stimulator, Ral guanine nucleotide exchange factor, RalGDS, RalGEFKIAA1308, RGDS, RGFFLJ20922 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 771-902 of human RALGDS (NP_001258705). CNSSSALPLYNQQVGDCCIIRVSLDVDNGNMYKSILVTSQDKAPAVIRKAMDKHNLEEEEPEDYELLQILSDDRKLKIPENANVFYAMNSTANYDFVLKKRTFTKGVKVKHGASSTLPRMKQKGLKIAKGIF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title