missing translation for 'onlineSavingsMsg'
Learn More

RanBP2 Antibody (2E1), Novus Biologicals™

Product Code. 18760274 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18760274 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18760274 Supplier Novus Biologicals Supplier No. NB10074480

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RanBP2 Monoclonal specifically detects RanBP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RanBP2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 20
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:1000, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunohistochemistry-Paraffin 1:100 - 1:1000
Gene Accession No. P49792
Gene Alias ANE1, E3 SUMO-protein ligase RanBP2, EC 5.2.1.8, Nuclear pore complex protein Nup358, nucleoporin 358, Nucleoporin Nup358, NUP358TRP2, P270, RAN binding protein 2, Ran-binding protein 2, RanBP2, transformation-related protein 2,358 kDa nucleoporin, TRP1
Gene Symbols RANBP2
Host Species Mouse
Immunogen Recombinant protein corresponding to 150 N-terminal residues of human Nup358.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5903
Test Specificity Nup358 (2E1)
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.