missing translation for 'onlineSavingsMsg'
Learn More

Ranitidine EP Impurity K, Mikromol™

Product Code. 30052510
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052510 25 mg 25mg
17952446 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30052510 Supplier Mikromol Supplier No. MM0220.090025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ranitidine EP Impurity K
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (EZ)-N-Methyl-1-(methylsulfanyl)-2-nitroethen-1-amine
CAS 61832-41-5
Analyte or Component Names (EZ)-N-Methyl-1-(methylsulfanyl)-2-nitroethen-1-amine
Shipping Condition Room Temperature
Molecular Formula C4 H8 N2 O2 S
Certifications/Compliance ISO 17025
Synonym (EZ)-N-Methyl-1-(methylsulphanyl)-2-nitroethen-1-amine, N-Methyl-1-methylthio-2-nitroethenamine, Nizatidine Imp. B (EP), Ranitidine Hydrochloride Imp. K (EP), Ranitidine Imp. K (EP), Ranitidine N-Methyl S-methyl, Ranitidine BP Impurity K, NMSM, Ph Eur Ranitidine Impurity K, Nizatidine Impurity B, Ranitidine Hydrochloride Impurity K, Ranitidine Impurity K
InChI Formula InChI=1S/C4H8N2O2S/c1-5-4(9-2)3-6(7)8/h3,5H,1-2H3
SMILES CNC(=C[N+](=O)[O-])SC
IUPAC Name N-methyl-1-methylsulfanyl-2-nitroethenamine
Molecular Weight (g/mol) 148.18
Quantity 25 mg
Formula Weight 148.0306 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.