missing translation for 'onlineSavingsMsg'
Learn More

RAP1A Antibody [mFluor Violet 450 SE], Novus Biologicals Biologicals™

Product Code. 30488026 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30488026 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30488026 Supplier Novus Biologicals Supplier No. NBP335089MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RAP1A Polyclonal antibody specifically detects RAP1A in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAP1A
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias C21KG, G-22K, GTP-binding protein smg p21A, KREV-1, KREV1Ras-related protein Krev-1, RAP1, RAP1A, member of RAS oncogene family, ras-related protein Rap-1A, RAS-related protein RAP1A, SMGP21
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 39-138 of human RAP1A (NP_002875.1).,, Sequence:, SYRKQVEVDCQQCMLEILDTAGTEQFTAMRDLYMKNGQGFALVYSITAQSTFNDLQDLREQILRVKDTEDVPMILVGNKCDLEDERVVGKEQGQNLARQW
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Endosome Markers
Primary or Secondary Primary
Gene ID (Entrez) 5906
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.