missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat VEGF164 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB564-500
This item is not returnable.
View return policy
Description
VEGF Monoclonal specifically detects VEGF in Rat samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| VEGF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7422 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, ELISA | |
| 123704 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| AAA41211.1 | |
| VEGFA | |
| Mouse myeloma cell line NS0-derived recombinant rat VEGF 164 Ala27-Arg190 Accession # AAA41211.1 | |
| 500 μg | |
| Primary | |
| Detects rat VEGF 164 in direct ELISAs and Western blots. Shows 50 - 100% cross-reactivity with recombinant mouse (rm) VEGF 120 rmVEGF 164 recombinant human (rh) VEGF 121 rhVEGF 165 and rhVEGF 189 and no cross-reactivity with rmVEGF 115 . Recognizes an epitope found between amino acids 78-120 of rmVEGF 120 . This segment is present in all VEGF isoforms except rmVEGF 115 . | |
| Rat | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction