missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat VEGF164 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
209.00 € - 2040.00 €
Specifications
| Antigen | VEGF |
|---|---|
| Clone | 123704 |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18745478
![]() |
R&D Systems
MAB564-SP |
25 μg |
209.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18703019
![]() |
R&D Systems
MAB564-100 |
100 μg |
512.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18723259
![]() |
R&D Systems
MAB564-500 |
500 μg |
2040.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VEGF Monoclonal specifically detects VEGF in Rat samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| VEGF | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Rat | |
| AAA41211.1 | |
| 7422 | |
| Mouse myeloma cell line NS0-derived recombinant rat VEGF 164 Ala27-Arg190 Accession # AAA41211.1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects rat VEGF 164 in direct ELISAs and Western blots. Shows 50 - 100% cross-reactivity with recombinant mouse (rm) VEGF 120 rmVEGF 164 recombinant human (rh) VEGF 121 rhVEGF 165 and rhVEGF 189 and no cross-reactivity with rmVEGF 115 . Recognizes an epitope found between amino acids 78-120 of rmVEGF 120 . This segment is present in all VEGF isoforms except rmVEGF 115 . |
| 123704 | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor | |
| VEGFA | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title