missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RB1, p Ser807 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-82177-0.025ML
This item is not returnable.
View return policy
Description
RB1 Polyclonal specifically detects RB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RB1 (p Ser807) | |
| Polyclonal | |
| Unconjugated | |
| PBS [pH 7.4] 150mM NaCl and 50% glycerol (without Mg2+ and Ca2+) with 0.02% Sodium Azide | |
| OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein | |
| Rabbit | |
| 110 kDa | |
| 0.025 mL | |
| Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors | |
| 5925 | |
| Human, Mouse, Rat | |
| IgG |
| Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
| P06400 | |
| RB1 | |
| The antiserum was produced against synthesized phosphopeptide derived from human Rb around the phosphorylation site of serine 807 (Y-I-SP-P-L). | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Rb (specific to Phospho-Ser807) detects endogenous levels of Rb only when phosphorylated at serine 807. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction