missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RB1, p Ser807 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
247.00 € - 406.00 €
Specifications
| Antigen | RB1 (p Ser807) |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18229892
|
Novus Biologicals
NB100-82177-0.025ML |
0.025 mL |
247.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18761294
|
Novus Biologicals
NB100-82177 |
0.05 mL |
406.00 €
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RB1 Polyclonal specifically detects RB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| RB1 (p Ser807) | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors | |
| P06400 | |
| 5925 | |
| IgG | |
| Affinity Purified | |
| 110 kDa |
| 1.0 mg/mL | |
| Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein | |
| RB1 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title