missing translation for 'onlineSavingsMsg'
Learn More

RBFOX3/NeuN Antibody (CL11892) [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30297069 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30297069 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30297069 Supplier Novus Biologicals Supplier No. NBP315124JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RBFOX3/NeuN Monoclonal antibody specifically detects RBFOX3/NeuN in Human,Mouse samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBFOX3/NeuN
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL11892
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias FLJ56884, FLJ58356, Fox-1 homolog C, FOX3, FOX-3, hexaribonucleotide binding protein 3, HRNBP3, NeuN, neuN antigen, neuronal nuclei, neuronal nuclei antigen, RBFOX3, RNA binding protein fox-1 homolog 3, RNA binding protein, fox-1 homolog (C. elegans) 3, RNA binding protein, fox-1 homolog 3
Host Species Mouse
Immunogen This antibody was developed using a synthetic peptide derived from A6NFN3, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 146713
Target Species Human, Mouse
Content And Storage Store at 4⪚C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.