missing translation for 'onlineSavingsMsg'
Learn More

RDM1 Antibody - BSA Free, R&D Systems™

Product Code. 18606322 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.01mL
0.02mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18606322 0.02 mL 0.02mL
18611452 0.1 mL 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18606322 Supplier Novus Biologicals Supplier No. NBP2952180.02ml
 Restricted Item
Estimated Shipment: 04-12-2026
Añadir a la cesta
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

RDM1 Polyclonal specifically detects RDM1 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Especificaciones

Antigen RDM1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:200-1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias MGC33977, RAD52 homolog B, RAD52 homolog B (S. cerevisiae), RAD52 motif 1, RAD52 motif-containing protein 1, RAD52B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 125-284 of human RDM1 (NP_663629.1). NGCSKRIIKLQELSDLEERENEDSMVPLPKQSLKFFCALEVVLPSCDCRSPGIGLVEEPMDKVEEGPLSFLMKRKTAQKLAIQKALSDAFQKLLIVVLESGKIAVEYRPSEDIVGVRCEEELHGLIQVPCSPWKQYGQEEEGYLSDFSLEEEEFRLPELD
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201299
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.