missing translation for 'onlineSavingsMsg'
Learn More

Reboxetine Mesilate, Mikromol™

Product Code. 18090400
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18090400 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18090400 Supplier Mikromol Supplier No. MM1613.00
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Reboxetine Mesilate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Reboxetine Mesilate
CAS 98769-84-7
Analyte or Component Names Reboxetine Mesilate
Shipping Condition Room Temperature
Molecular Formula C19 H23 N O3 . C H4 O3 S
Certifications/Compliance ISO 17025
Synonym Reboxetine Mesilate, Morpholine, 2-[(R)-(2-ethoxyphenoxy)phenylmethyl]-, (2R)-rel-, methanesulfonate (9CI), Davedax, Edronax, FCE 20124, PNU 155905E, PNU 155950E, Reboxetine mesilate, Reboxetine mesylate, Morpholine, 2-[(R)-(2-ethoxyphenoxy)phenylmethyl]-, (2R)-rel-, methanesulfonate (1:1), Morpholine, 2-[(2-ethoxyphenoxy)phenylmethyl]-, (R*,R*)-(+/-)-, methanesulfonate, (+/-) Reboxetine mesylate
InChI Formula InChI=1S/C19H23NO3.CH4O3S/c1-2-21-16-10-6-7-11-17(16)23-19(15-8-4-3-5-9-15)18-14-20-12-13-22-18;1-5(2,3)4/h3-11,18-20H,2,12-14H2,1H3;1H3,(H,2,3,4)/t18-,19-;/m1./s1
SMILES CCOc1ccccc1O[C@@H]([C@H]2CNCCO2)c3ccccc3.CS(=O)(=O)O
IUPAC Name (2R)-2-[(R)-(2-ethoxyphenoxy)-phenylmethyl]morpholine;methanesulfonic acid
Molecular Weight (g/mol) 409.5
Quantity 50 mg
Formula Weight 409.1559 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.