Learn More
Invitrogen™ RECA-1 Monoclonal Antibody (HIS52)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA181510
Description
This antibody does not react with chicken, goat, guinea pig, mouse, pig, rabbit or sheep. Mouse anti Rat RECA-1 antibody, clone HIS52 recognizes RECA-1, a cell surface antigen which is expressed by all rat endothelial cells. Mouse anti Rat RECA-1 antibody, clone HIS52 recognizes RECA-1, a cell surface antigen which is expressed by all rat endothelial cells.
RECA-1 is a cell surface antigen which is expressed by all rat endothelial cells. The endothelium is the thin layer of cells that line the interior surface of blood vessels, forming an interface between circulating blood in the lumen and the rest of the vessel wall. Endothelial cells line the entire circulatory system, from the heart to the smallest capillary.
Specifications
| RECA-1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with <0.1% sodium azide | |
| Mouse | |
| Protein G | |
| RUO | |
| Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Frozen), Immunocytochemistry | |
| HIS52 | |
| Unconjugated | |
| Reca1 | |
| Stromal cells from rat lymph node. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.