missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RECA-1 Monoclonal Antibody (HIS52)
GREENER_CHOICE

Mouse Monoclonal Antibody

522.00 € - 730.00 €

Specifications

Antigen RECA-1
Clone HIS52
Concentration 0.5 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications Immunohistochemistry (Frozen), Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11545412
View Documents
Invitrogen™
MA180181
250 μg
730.00 €
250µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11510013
GREENER_CHOICE
View Documents
Invitrogen™
MA181510
100 μg
522.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody does not react with chicken, goat, guinea pig, mouse, pig, rabbit or sheep. Mouse anti Rat RECA-1 antibody, clone HIS52 recognizes RECA-1, a cell surface antigen which is expressed by all rat endothelial cells. Mouse anti Rat RECA-1 antibody, clone HIS52 recognizes RECA-1, a cell surface antigen which is expressed by all rat endothelial cells.

RECA-1 is a cell surface antigen which is expressed by all rat endothelial cells. The endothelium is the thin layer of cells that line the interior surface of blood vessels, forming an interface between circulating blood in the lumen and the rest of the vessel wall. Endothelial cells line the entire circulatory system, from the heart to the smallest capillary.
Specifications

Specifications

RECA-1
0.5 mg/mL
Immunohistochemistry (Frozen), Immunocytochemistry
Unconjugated
Mouse
RUO
PBS with <0.1% sodium azide
Stromal cells from rat lymph node.
Antibody
HIS52
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Monoclonal
Liquid
IgG1
Rat
Reca1
Primary
Protein G
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.